N-[4-(Aminomethyl)benzyl]-N,N--dimethylamine 98%

Lieferant: Apollo Scientific

Synonyme: 1-(4-(Aminomethyl)phenyl)-N,N-dimethylmethanamine, 4-Dimethylaminomethylbenzylamine

OR953945-1G
APOSOR953945-1GEA 140 EUR
APOSOR953945-1G
N-[4-(Aminomethyl)benzyl]-N,N--dimethylamine 98%
N-[4-(Aminomethyl)benzyl]-N,N--dimethylamine
Formel: C₁₀H₁₆N₂
Molecular Weight: 164.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04107485
CAS-Nummer: 34490-85-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR