5-(Aminomethyl)-N,N-dimethyl-2-pyridinamine 98%

Lieferant: Apollo Scientific

OR953983-1G OR953983-250MG OR953983-5G
APOSOR953983-1GEA 270 EUR
APOSOR953983-1G APOSOR953983-250MG APOSOR953983-5G
5-(Aminomethyl)-N,N-dimethyl-2-pyridinamine 98%
5-(Aminomethyl)-N,N-dimethyl-2-pyridinamine
Formel: C₈H₁₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08272104
CAS-Nummer: 354824-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR