2-(Ethylthio)benzoic acid 95%

Lieferant: Apollo Scientific

OR954047-1G OR954047-250MG
APOSOR954047-1GEA 435 EUR
APOSOR954047-1G APOSOR954047-250MG
2-(Ethylthio)benzoic acid 95%
2-(Ethylthio)benzoic acid
Formel: C₉H₁₀O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00094046
CAS-Nummer: 21101-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR