Methyl-2-pyridinylacetat 95%

Lieferant: Apollo Scientific

Synonyme: 2-Pyridineacetic Acid Methyl Ester, Methyl 2-Pyridineacetate, Methyl 2-(pyridin-2-yl)acetate, 2-Pyridylacetic Acid Methyl Ester

OR954071-25G
APOSOR954071-25GEA 98.6 EUR
APOSOR954071-25G
Methyl-2-pyridinylacetat 95%
Methyl-2-pyridinylacetat
Formel: C₈H₉NO₂
Siedepunkt: 137 °C
Dichte: 1,13 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006358
CAS-Nummer: 1658-42-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR