4-(1-Methyl-5-imidazolyl)phenol 95%
Lieferant: Apollo Scientific
OR954116-1G
APOSOR954116-1GEA
415
EUR
APOSOR954116-1G
4-(1-Methyl-5-imidazolyl)phenol 95%
4-(1-Methyl-5-imidazolyl)phenol
Formel:
C₁₀H₁₀N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD18375274 CAS-Nummer: 74730-75-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...