2-(Dimethylamino)pyrimidine-5-carbaldehyde 95%

Lieferant: Apollo Scientific

OR954195-1G
APOSOR954195-1GEA 0 EUR
APOSOR954195-1G
2-(Dimethylamino)pyrimidine-5-carbaldehyde 95%
2-(Dimethylamino)pyrimidine-5-carbaldehyde
Formel: C₇H₉N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD03446733
CAS-Nummer: 55551-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR