2-Chloroquinolin-6-ol 98%

Lieferant: Apollo Scientific

OR954239-5G
APOSOR954239-5GEA 121 EUR
APOSOR954239-5G
2-Chloroquinolin-6-ol 98%
2-Chloroquinolin-6-ol
Formel: C₉H₆ClNO
Molecular Weight: 179.603 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12405093
CAS-Nummer: 577967-89-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR