1,2-Dimethylbenzimidazole 98%
Lieferant: Apollo Scientific
OR954313-1G
OR954313-5G
APOSOR954313-1GEA
72.5
EUR
APOSOR954313-1G
APOSOR954313-5G
1,2-Dimethylbenzimidazole 98%
1,2-Dimethylbenzimidazole
Formel:
C₉H₁₀N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022840 CAS-Nummer: 2876-08-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...