4,4,6-Trimethyl-1,3-dihydroquinolin-2-one 98%

Lieferant: Apollo Scientific
Achtung

OR954461-1G
APOSOR954461-1GEA 69.6 EUR
APOSOR954461-1G
4,4,6-Trimethyl-1,3-dihydroquinolin-2-one 98%
4,4,6-Trimethyl-1,3-dihydroquinolin-2-one
Formel: C₁₂H₁₅NO
Molecular Weight: 189,254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12913845
CAS-Nummer: 181122-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR