5-Acetamido-2-chloropyridine 97%

Lieferant: Apollo Scientific

OR954492-1G
APOSOR954492-1GEA 574 EUR
APOSOR954492-1G
5-Acetamido-2-chloropyridine 97%
5-Acetamido-2-chloropyridine
Formel: C₇H₇ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD20277888
CAS-Nummer: 29958-18-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR