5-(4-Methoxyphenyl)-3H-1,3,4-oxadiazol-2-one 98%

Lieferant: Apollo Scientific

OR954532-1G
APOSOR954532-1GEA 418 EUR
APOSOR954532-1G
5-(4-Methoxyphenyl)-3H-1,3,4-oxadiazol-2-one 98%
5-(4-Methoxyphenyl)-3H-1,3,4-oxadiazol-2-one
Formel: C₉H₈N₂O₃
Molecular Weight: 192,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669523
CAS-Nummer: 41125-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR