2-(3-Methoxyphenyl)-1,3,4-oxadiazole 98%

Lieferant: Apollo Scientific

OR954533-1G
APOSOR954533-1GEA 400 EUR
APOSOR954533-1G
2-(3-Methoxyphenyl)-1,3,4-oxadiazole 98%
2-(3-Methoxyphenyl)-1,3,4-oxadiazole
Formel: C₉H₈N₂O₂
Molecular Weight: 176,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09755524
CAS-Nummer: 5378-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR