3-Ethoxy-N-methyl-4-nitroaniline 98%
Lieferant: Apollo Scientific
OR954634-1G
APOSOR954634-1GEA
400
EUR
APOSOR954634-1G
3-Ethoxy-N-methyl-4-nitroaniline 98%
3-Ethoxy-N-methyl-4-nitroaniline
Formel:
C₉H₁₂N₂O₃ Molecular Weight: 196,203 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18861025 CAS-Nummer: 1339134-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...