2-Methanesulphonyl-5-methylaniline 95%

Lieferant: Apollo Scientific

OR954645-1G OR954645-5G
APOSOR954645-1GEA 219 EUR
APOSOR954645-1G APOSOR954645-5G
2-Methanesulphonyl-5-methylaniline 95%
2-Methanesulphonyl-5-methylaniline
Formel: C₈H₁₁NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD11645433
CAS-Nummer: 876494-64-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR