3-Cyan-5-nitrophenylboronsäure 96+%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Cyano-5-nitrophenyl)boronic acid

OR9547-1G
APOSOR9547-1GEA 282 EUR
APOSOR9547-1G
3-Cyan-5-nitrophenylboronsäure 96+%
3-Cyan-5-nitrophenylboronsäure
Formel: C₇H₅BN₂O₄
Molecular Weight: 191,936 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03411554
CAS-Nummer: 913835-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR