Biphenyl-3-ol 98%

Lieferant: Apollo Scientific

Synonyme: 3-Hydroxybiphenyl, 3-Hydroxydiphenyl, 1-Hydroxy-3-phenylbenzol, 3-Phenylphenol

OR954760-25G
APOSOR954760-25GEA 95.7 EUR
APOSOR954760-25G
Biphenyl-3-ol 98%
Biphenyl-3-ol
Formel: C₁₂H₁₀O
Molecular Weight: 170.21 g/mol
Siedepunkt: 334
Schmelzpunkt: 78
Lagertemperatur: Raumtemperatur
MDL: MFCD00002294
CAS-Nummer: 580-51-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR