3-(2-Methylphenyl)phenol 96%

Lieferant: Apollo Scientific

Synonyme: 2'-Methyl-[1,1'-biphenyl]-3-ol

OR954764-1G
APOSOR954764-1GEA 266 EUR
APOSOR954764-1G
3-(2-Methylphenyl)phenol 96%
3-(2-Methylphenyl)phenol
Formel: C₁₃H₁₂O
Molecular Weight: 184,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06802319
CAS-Nummer: 71965-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR