2-(3-Methylphenyl)phenol 98%
Lieferant: Apollo Scientific
OR954766-1G
APOSOR954766-1GEA
0
EUR
APOSOR954766-1G
2-(3-Methylphenyl)phenol 98%
2-(3-Methylphenyl)phenol
Formel:
C₁₃H₁₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD06802436 CAS-Nummer: 214268-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...