2-(4-Methylphenyl)phenol 96%
Lieferant: Apollo Scientific
OR954769-1G
APOSOR954769-1GEA
364
EUR
APOSOR954769-1G
2-(4-Methylphenyl)phenol 96%
2-(4-Methylphenyl)phenol
Formel:
C₁₃H₁₂O Molecular Weight: 184,234 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06802423 CAS-Nummer: 101043-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...