4-(2-Hydroxyphenyl)phenol 98%

Lieferant: Apollo Scientific

Synonyme: [1,1'-Biphenyl]-2,4'-diol

OR954775-1G
APOSOR954775-1GEA 401 EUR
APOSOR954775-1G
4-(2-Hydroxyphenyl)phenol 98%
4-(2-Hydroxyphenyl)phenol
Formel: C₁₂H₁₀O₂
Molecular Weight: 186.207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11895228
CAS-Nummer: 611-62-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR