2-(4-Cyanophenyl)phenol 96%
Lieferant: Apollo Scientific
OR954779-1G
APOSOR954779-1GEA
364
EUR
APOSOR954779-1G
2-(4-Cyanophenyl)phenol 96%
2-(4-Cyanophenyl)phenol
Formel:
C₁₃H₉NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD06802201 CAS-Nummer: 127703-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...