4-(2-Formylphenyl)phenol 95%

Lieferant: Apollo Scientific

OR954780-1G OR954780-250MG
APOSOR954780-1GEA 538 EUR
APOSOR954780-1G APOSOR954780-250MG
4-(2-Formylphenyl)phenol 95%
4-(2-Formylphenyl)phenol
Formel: C₁₃H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04117373
CAS-Nummer: 400747-55-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR