3-(3-Formylphenyl)phenol 96%

Lieferant: Apollo Scientific

OR954781-1G
APOSOR954781-1GEA 364 EUR
APOSOR954781-1G
3-(3-Formylphenyl)phenol 96%
3-(3-Formylphenyl)phenol
Formel: C₁₃H₁₀O₂
Molecular Weight: 198,217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04117377
CAS-Nummer: 400745-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR