4-(3-Formylphenyl)phenol 98%

Lieferant: Apollo Scientific

OR954782-1G
APOSOR954782-1GEA 435 EUR
APOSOR954782-1G
4-(3-Formylphenyl)phenol 98%
4-(3-Formylphenyl)phenol
Formel: C₁₃H₁₀O₂
Molecular Weight: 198,217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04117372
CAS-Nummer: 283147-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR