2-(4-Formylphenyl)phenol 95%

Lieferant: Apollo Scientific

OR954783-1G
APOSOR954783-1GEA 632 EUR
APOSOR954783-1G
2-(4-Formylphenyl)phenol 95%
2-(4-Formylphenyl)phenol
Formel: C₁₃H₁₀O₂
Molecular Weight: 198.217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06801753
CAS-Nummer: 400744-38-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR