2-Methyl-5-phenylphenol 95%

Lieferant: Apollo Scientific

OR954882-1G
APOSOR954882-1GEA 451 EUR
APOSOR954882-1G
2-Methyl-5-phenylphenol 95%
2-Methyl-5-phenylphenol
Formel: C₁₃H₁₂O
Molecular Weight: 184,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03265274
CAS-Nummer: 105902-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR