2-Methyl-5-(thiophen-3-yl)phenol 96%

Lieferant: Apollo Scientific

OR954885-1G
APOSOR954885-1GEA 364 EUR
APOSOR954885-1G
2-Methyl-5-(thiophen-3-yl)phenol 96%
2-Methyl-5-(thiophen-3-yl)phenol
Formel: C₁₁H₁₀OS
Molecular Weight: 190,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18313076
CAS-Nummer: 1261985-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR