2-Bromo-5-(hydroxymethyl)benzonitrile 98%

Lieferant: Apollo Scientific
Achtung

OR955151-250MG
APOSOR955151-250MGEA 133 EUR
APOSOR955151-250MG
2-Bromo-5-(hydroxymethyl)benzonitrile 98%
2-Bromo-5-(hydroxymethyl)benzonitrile
Formel: C₈H₆BrNO
Molecular Weight: 212,043 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18390752
CAS-Nummer: 1261609-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR