3-Formylbenzamide 95%

Lieferant: Apollo Scientific

OR955186-1G
APOSOR955186-1GEA 98.6 EUR
APOSOR955186-1G
3-Formylbenzamide 95%
3-Formylbenzamide
Formel: C₈H₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09758970
CAS-Nummer: 126534-87-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR