(3,5-Dimethylpyridin-2-yl)methanol 95%

Lieferant: Apollo Scientific

OR955234-1G
APOSOR955234-1GEA 68.3 EUR
APOSOR955234-1G
(3,5-Dimethylpyridin-2-yl)methanol 95%
(3,5-Dimethylpyridin-2-yl)methanol
Formel: C₈H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09839185
CAS-Nummer: 202932-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR