5-(3-Methylphenyl)pyridin-2-amine 95%
Lieferant: Apollo Scientific
OR955283-1G
APOSOR955283-1GEA
364
EUR
APOSOR955283-1G
5-(3-Methylphenyl)pyridin-2-amine 95%
5-(3-Methylphenyl)pyridin-2-amine
Formel:
C₁₂H₁₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06802146 CAS-Nummer: 893738-40-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...