4'-Chloro-biphenyl-4-acetic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-(4'-Chloro-[1,1'-biphenyl]-4-yl)acetic acid

OR955330-1G
APOSOR955330-1GEA 149 EUR
APOSOR955330-1G
4'-Chloro-biphenyl-4-acetic acid 97%
4'-Chloro-biphenyl-4-acetic acid
Formel: C₁₄H₁₁ClO₂
Molecular Weight: 246.689 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03426507
CAS-Nummer: 5525-72-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR