3-(4-Methylphenyl)pyridine-2-carbonitrile 98%

Lieferant: Apollo Scientific

OR955383-1G
APOSOR955383-1GEA 502 EUR
APOSOR955383-1G
3-(4-Methylphenyl)pyridine-2-carbonitrile 98%
3-(4-Methylphenyl)pyridine-2-carbonitrile
Formel: C₁₃H₁₀N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22586704
CAS-Nummer: 143425-48-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR