3-(2-Methylphenyl)pyridine-2-carbonitrile 98%
Lieferant: Apollo Scientific
OR955384-1G
APOSOR955384-1GEA
435
EUR
APOSOR955384-1G
3-(2-Methylphenyl)pyridine-2-carbonitrile 98%
3-(2-Methylphenyl)pyridine-2-carbonitrile
Formel:
C₁₃H₁₀N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22586705 CAS-Nummer: 1415819-89-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...