6-Phenyl-2-(pyrazin-2-yl)pyrimidin-4-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Hydroxy-6-phenyl-1,3-diazin-2-yl)pyrazine, 4-Hydroxy-6-phenyl-2-(pyrazin-2-yl)pyrimidine

OR9554-50MG
APOSOR9554-50MGEA 21.9 EUR
APOSOR9554-50MG
6-Phenyl-2-(pyrazin-2-yl)pyrimidin-4-ol
6-Phenyl-2-(pyrazin-2-yl)pyrimidin-4-ol
Formel: C₁₄H₁₀N₄O
Molecular Weight: 250,255 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665788
CAS-Nummer: 874605-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR