3-Methoxy-5-phenylaniline 98%

Lieferant: Apollo Scientific

OR955414-1G
APOSOR955414-1GEA 632 EUR
APOSOR955414-1G
3-Methoxy-5-phenylaniline 98%
3-Methoxy-5-phenylaniline
Formel: C₁₃H₁₃NO
Molecular Weight: 199,248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20702469
CAS-Nummer: 383870-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR