Methyl-2-amino-6-chlorbenzoat 98%

Lieferant: Apollo Scientific

OR955428-1G
APOSOR955428-1GEA 74.1 EUR
APOSOR955428-1G
Methyl-2-amino-6-chlorbenzoat 98%
Methyl-2-amino-6-chlorbenzoat
Formel: C₈H₈ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16036437
CAS-Nummer: 41632-04-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR