D-Phenylalanine methyl ester 95%

Lieferant: Apollo Scientific

OR955432-1G
APOSOR955432-1GEA 435 EUR
APOSOR955432-1G
D-Phenylalanine methyl ester 95%
D-Phenylalanine methyl ester
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179.216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07786601
CAS-Nummer: 21685-51-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR