3-Chloro-2-methyl-benzoic acid methyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-chloro-2-methylbenzoate

OR955456-5G
APOSOR955456-5GEA 169 EUR
APOSOR955456-5G
3-Chloro-2-methyl-benzoic acid methyl ester 95%
3-Chloro-2-methyl-benzoic acid methyl ester
Formel: C₉H₉ClO₂
Molecular Weight: 184.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00230148
CAS-Nummer: 99586-84-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR