Methyl-6-hydroxypyridine-2-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 6-oxo-1,6-dihydropyridine-2-carboxylate

OR955459-5G
APOSOR955459-5GEA 82.8 EUR
APOSOR955459-5G
Methyl-6-hydroxypyridine-2-carboxylate 98%
Methyl-6-hydroxypyridine-2-carboxylate
Formel: C₇H₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD14698054
CAS-Nummer: 30062-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR