methyl isoxazole-3-carboxylate 95%
Lieferant: Apollo Scientific
OR955465-1G
APOSOR955465-1GEA
435
EUR
APOSOR955465-1G
methyl isoxazole-3-carboxylate 95%
methyl isoxazole-3-carboxylate
Formel:
C₅H₅NO₃ Molecular Weight: 127,098 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11053758 CAS-Nummer: 25742-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...