2,4,6-Trimethyl-benzeneacetic acid methyl ester 95%

Lieferant: Apollo Scientific

OR955536-1G
APOSOR955536-1GEA 632 EUR
APOSOR955536-1G
2,4,6-Trimethyl-benzeneacetic acid methyl ester 95%
2,4,6-Trimethyl-benzeneacetic acid methyl ester
Formel: C₁₂H₁₆O₂
Molecular Weight: 192,254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12024951
CAS-Nummer: 41841-19-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR