3-(2-Pyridinyl)-2-propenoic acid methyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-(pyridin-2-yl)acrylate

OR955550-1G
APOSOR955550-1GEA 208 EUR
APOSOR955550-1G
3-(2-Pyridinyl)-2-propenoic acid methyl ester 95%
3-(2-Pyridinyl)-2-propenoic acid methyl ester
Formel: C₉H₉NO₂
Molecular Weight: 163,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18803084
CAS-Nummer: 50610-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR