Methyl 1,2,3,4-tetrahydronaphthalene-1-carboxylate 98%

Lieferant: Apollo Scientific

OR955554-1G
APOSOR955554-1GEA 435 EUR
APOSOR955554-1G
Methyl 1,2,3,4-tetrahydronaphthalene-1-carboxylate 98%
Methyl 1,2,3,4-tetrahydronaphthalene-1-carboxylate
Formel: C₁₂H₁₄O₂
Molecular Weight: 190,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11816647
CAS-Nummer: 17502-86-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR