Methyl (2r)-2-(methylamino)propanoate hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: (R)-Methyl 2-(methylamino)propanoate hydrochloride

OR955570-1G
APOSOR955570-1GEA 435 EUR
APOSOR955570-1G
Methyl (2r)-2-(methylamino)propanoate hydrochloride 97%
Methyl (2r)-2-(methylamino)propanoate hydrochloride
Formel: C₅H₁₁NO₂·ClH
Molecular Weight: 153,607 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973243
CAS-Nummer: 19914-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR