Methyl quinoline-4-carboxylate 97%
Lieferant: Apollo Scientific
OR955585-1G
APOSOR955585-1GEA
271
EUR
APOSOR955585-1G
Methyl quinoline-4-carboxylate 97%
Methyl quinoline-4-carboxylate
Formel:
C₁₁H₉NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01366551 CAS-Nummer: 21233-61-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...