Methyl (E)-3-(2-methylpyrazol-3-yl)prop-2-enoate 95%

Lieferant: Apollo Scientific

OR955588-1G
APOSOR955588-1GEA 632 EUR
APOSOR955588-1G
Methyl (E)-3-(2-methylpyrazol-3-yl)prop-2-enoate 95%
Methyl (E)-3-(2-methylpyrazol-3-yl)prop-2-enoate
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166.177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23716020
CAS-Nummer: 1563651-10-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR