Methyl-3-(4-pyridyl)acrylate 95%

Lieferant: Apollo Scientific

OR955608-1G
APOSOR955608-1GEA 151 EUR
APOSOR955608-1G
Methyl-3-(4-pyridyl)acrylate 95%
Methyl-3-(4-pyridyl)acrylate
Formel: C₉H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00023636
CAS-Nummer: 7340-34-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR