Methyl 2-amino-3,4-dimethylbenzoate 95%

Lieferant: Apollo Scientific

OR955614-1G
APOSOR955614-1GEA 632 EUR
APOSOR955614-1G
Methyl 2-amino-3,4-dimethylbenzoate 95%
Methyl 2-amino-3,4-dimethylbenzoate
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12159829
CAS-Nummer: 31928-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR