(R)-Methyl 2-methoxy-2-phenylacetate 95%
Lieferant: Apollo Scientific
OR955653-1G
APOSOR955653-1GEA
435
EUR
APOSOR955653-1G
(R)-Methyl 2-methoxy-2-phenylacetate 95%
(R)-Methyl 2-methoxy-2-phenylacetate
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180.2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16879060 CAS-Nummer: 32174-46-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...